Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Hexylpyridinium Hexafluorophosphate 98.0+%, TCI America™
SDP

Catalog No. H142425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H142425G 25 g
H14245G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H142425G Supplier TCI America Supplier No. H142425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 797789-00-5
Molecular Formula C11H18F6NP
Molecular Weight (g/mol) 309.236
MDL Number MFCD03093298
InChI Key DKRTWJPQQYAIHH-UHFFFAOYSA-N
Synonym 1-hexylpyridinium hexafluorophosphate, 1-hexylpyridin-1-ium hexafluorophosphate v, 1-hexylpyridin-1-ium hexafluorophosphate, dsstox_cid_27912, dsstox_rid_82665, dsstox_gsid_47936, ksc376g7d, n-hexylpyridinium hexafluorophosphate
PubChem CID 2734177
IUPAC Name 1-hexylpyridin-1-ium;hexafluorophosphate
SMILES CCCCCC[N+]1=CC=CC=C1.F[P-](F)(F)(F)(F)F

Specifications

Melting Point 48°C
Color White
Quantity 25 g
Formula Weight 309.24
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Hexylpyridinium Hexafluorophosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.