Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Hydroxy-4-nitroanthraquinone 97.0+%, TCI America™
SDP

Catalog No. H076525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H076525G 25 g
H07655G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H076525G Supplier TCI America Supplier No. H076525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 81-65-2
Molecular Formula C14H7NO5
Molecular Weight (g/mol) 269.212
MDL Number MFCD00142642
InChI Key XXUOKOYLVBHBCG-UHFFFAOYSA-N
PubChem CID 3490678
IUPAC Name 1-hydroxy-4-nitroanthracene-9,10-dione
SMILES C1=CC=C2C(=C1)C(=O)C3=C(C=CC(=C3C2=O)O)[N+](=O)[O-]

Specifications

Melting Point 272°C
Quantity 25 g
Formula Weight 269.21
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Hydroxy-4-nitroanthraquinone
EINECSNumber (4)-0735
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.