Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Iodo-2-methylpropane, 97%, stabilized

Catalog No. AC223741000
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC223741000 100 mL Glass bottle
AC223740250 25 mL Glass bottle
AC223745000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC223741000 Supplier Thermo Scientific Chemicals Supplier No. 223741000
Add to Cart

CAS: 513-38-2 | C4H9I | 184.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 513-38-2
Molecular Formula C4H9I
Molecular Weight (g/mol) 184.02
MDL Number MFCD00001084
InChI Key BTUGGGLMQBJCBN-UHFFFAOYSA-N
Synonym isobutyl iodide, propane, 1-iodo-2-methyl, isobutyljodid, isobutyliodide, 2-methylpropyl iodide, primary isobutyl iodide, isobutyljodid czech, 1-jod-2-methylpropan, 1-jod-2-methylpropan czech, 1-iodo-2-methyl-propane
PubChem CID 10556
IUPAC Name 1-iodo-2-methylpropane
SMILES CC(C)CI

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -93.0°C
Color Colorless to Orange
Density 1.5900g/mL
Boiling Point 120.0°C to 121.0°C
Flash Point 12°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4948 to 1.4968
Linear Formula (CH3)2CHCH2I
Quantity 100 mL
Merck Index 15,5189
Specific Gravity 1.59
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with alcohol and ether
Stabilizer copper
Formula Weight 184.02
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Iodo-2-methylpropane, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.
Harmful if inhaled.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 208-160-5
RTECSNumber TZ4250000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.