Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Iodo-2-nitrobenzene, 97%, Thermo Scientific Chemicals

Catalog No. AC159451000
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC159451000 100 g
AC159450250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC159451000 Supplier Thermo Scientific Chemicals Supplier No. 159451000

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 609-73-4
Molecular Formula C6H4INO2
Molecular Weight (g/mol) 249.01
MDL Number MFCD00007088
InChI Key JXMZUNPWVXQADG-UHFFFAOYSA-N
Synonym 2-iodonitrobenzene, o-iodonitrobenzene, o-nitroiodobenzene, benzene, 1-iodo-2-nitro, 2-nitroiodobenzene, 2-iodo-1-nitrobenzene, 1-iodo-2-nitro-benzene, 2-iodo-nitrobenzene, 2-nitro-iodobenzene, benzene, iodonitro
PubChem CID 69115
IUPAC Name 1-iodo-2-nitrobenzene
SMILES [O-][N+](=O)C1=CC=CC=C1I

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 47°C to 52°C
Color Brown-Yellow to Green-Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Linear Formula IC6H4NO2
Quantity 100 g
Beilstein 05, 252
Formula Weight 249.01
Percent Purity ≥96.0%
Physical Form Granular Powder
Chemical Name or Material 1-Iodo-2-nitrobenzene, 97%
Show More Show Less
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 210-200-1
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.