Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Iodo-2-(trifluoromethoxy)benzene 98.0+%, TCI America™
SDP

Catalog No. T215225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T215225G 25 g
T21525G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T215225G Supplier TCI America Supplier No. T215225G

Chemical Identifiers

CAS 175278-00-9
Molecular Formula C7H4F3IO
Molecular Weight (g/mol) 288.01
MDL Number MFCD00042410
InChI Key GYBMJVZOZTVDKS-UHFFFAOYSA-N
Synonym 1-iodo-2-trifluoromethoxy benzene, 2-trifluoromethoxy iodobenzene, benzene, 1-iodo-2-trifluoromethoxy, 2-iodo trifluoromethoxy benzene, o-iodotrifluoromethoxybenzene, o-trifluoromethoxy iodobenzene, alpha,alpha,alpha-trifluoro-2-iodoanisole, pubchem1077, acmc-1ca3h, ksc494o5n
PubChem CID 2777292
IUPAC Name 1-iodo-2-(trifluoromethoxy)benzene
SMILES FC(F)(F)OC1=CC=CC=C1I

Specifications

Color Yellow
Boiling Point 65°C
Quantity 25 g
Formula Weight 288.01
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Iodo-2-(trifluoromethoxy)benzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.