Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Iodo-4-nitrobenzene 99.0+%, TCI America™
SDP

Catalog No. I0064100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0064100G 100 g
I006425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I0064100G Supplier TCI America Supplier No. I0064100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 636-98-6
Molecular Formula C6H4INO2
Molecular Weight (g/mol) 249.007
MDL Number MFCD00007299
InChI Key SCCCFNJTCDSLCY-UHFFFAOYSA-N
Synonym benzene, 1-iodo-4-nitro, p-iodonitrobenzene, 4-iodonitrobenzene, p-nitroiodobenzene, 4-nitroiodobenzene, 4-iodo-nitrobenzene, p-nitrophenyl iodide, 1-iodo-4-nitro-benzene, 4-iodo-1-nitrobenzene, 4-nitro-1-iodobenzene
PubChem CID 12495
IUPAC Name 1-iodo-4-nitrobenzene
SMILES C1=CC(=CC=C1[N+](=O)[O-])I

Specifications

Melting Point 173°C
Quantity 100 g
Formula Weight 249.01
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Iodo-4-nitrobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.