Learn More
1-Iododecane, 98%, stab. with copper
Description
1-Iododecane is used as a precursor to prepare 9-nonadecanone through palladium-catalyzed carbonylative cross-coupling reaction with 9-octyl-9-borabicyclo[3.3.l]nonane. It is also used as an intermediate in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1-Iododecane is used as a precursor to prepare 9-nonadecanone through palladium-catalyzed carbonylative cross-coupling reaction with 9-octyl-9-borabicyclo[3.3.l]nonane. It is also used as an intermediate in organic synthesis.
Solubility
Immiscible with water.
Notes
Light sensitive. Incompatible with strong oxidizing agents and strong bases. Keep the container tightly closed.
Specifications
Specifications
| Melting Point | -16°C |
| Density | 1.257 |
| Boiling Point | 264°C |
| Flash Point | 96°C (204°F) |
| Refractive Index | 1.485 |
| Linear Formula | CH3(CH2)9I |
| Quantity | 50 g |
| Beilstein | 1735228 |
| Sensitivity | Light sensitive |
| Synonym | decyl iodide, decane, 1-iodo, n-decyl iodide, iododecane, 1-iodanyldecane, 1-decyl iodide, 1-iodo-decane, 1-iod0decane, decane,1-iodo, pubchem3942 |
| Show More |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319 Causes skin irritation. Causes serious eye irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 218-104-1 |
| RTECSNumber | HD8705000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.