Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Iodopropane (stabilized with Copper chip) 98.0+%, TCI America™
SDP

Catalog No. I0068100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0068100G 100 g
I006825G 25 g
I0068500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I0068100G Supplier TCI America Supplier No. I0068100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 107-08-4
Molecular Formula C3H7I
Molecular Weight (g/mol) 169.99
MDL Number MFCD00001095
InChI Key FMKOJHQHASLBPH-UHFFFAOYSA-N
Synonym propyl iodide, n-propyl iodide, propane, 1-iodo, propane, iodo, 1-iodo-propane, 1-jodpropan, iodopropane, unii-nd3629ke2k, propyliodide, iodopropanes
PubChem CID 33643
IUPAC Name 2-iodopropane
SMILES CC(C)I

Specifications

Color Yellow
Boiling Point 102°C
Quantity 100 g
UN Number 2392
Formula Weight 169.99
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Iodopropane (stabilized with Copper chip)
RTECSNumber TZ4100000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.