Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Isopropylnaphthalene 85.0+%, TCI America™
SDP

Catalog No. I05781G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I05781G 1 g
I05785G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I05781G Supplier TCI America Supplier No. I05781G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6158-45-8
Molecular Formula C13H14
Molecular Weight (g/mol) 170.255
MDL Number MFCD00061059
InChI Key PMPBFICDXLLSRM-UHFFFAOYSA-N
PubChem CID 22535
IUPAC Name 1-propan-2-ylnaphthalene
SMILES CC(C)C1=CC=CC2=CC=CC=C21

Specifications

Melting Point -16°C
Color Yellow
Boiling Point 268°C
Quantity 1 g
Formula Weight 170.26
Percent Purity ≥85.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Isopropylnaphthalene
EINECSNumber (4)-0961
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.