Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Isopropylpiperazine 98.0+%, TCI America™
SDP

Catalog No. I074525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I074525G 25 g
I07455G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I074525G Supplier TCI America Supplier No. I074525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4318-42-7
Molecular Formula C7H16N2
Molecular Weight (g/mol) 128.219
MDL Number MFCD00167971
InChI Key WHKWMTXTYKVFLK-UHFFFAOYSA-N
Synonym 1-isopropylpiperazine, n-isopropylpiperazine, 1-propan-2-yl piperazine, piperazine, 1-isopropyl, isopropyl piperazine, 1-isopropyl-piperazine, 1-1-methylethyl piperazine, n-isopropyl piperazine, 1-isopropyl piperazine, piperazine, 1-1-methylethyl
PubChem CID 78013
IUPAC Name 1-propan-2-ylpiperazine
SMILES CC(C)N1CCNCC1

Specifications

Color Yellow
Boiling Point 178°C
Quantity 25 g
UN Number 1992
Formula Weight 128.22
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-Isopropylpiperazine
RTECSNumber TM0860000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.