Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Mercaptoadamantane, 98%

Catalog No. AAH5895803
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5895803 1 g
AAH58958MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5895803 Supplier Thermo Scientific Chemicals Supplier No. H5895803
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 34301-54-7 | C10H16S | 168.298 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34301-54-7
Molecular Formula C10H16S
Molecular Weight (g/mol) 168.298
MDL Number MFCD00213530
InChI Key ADJJLNODXLXTIH-UHFFFAOYSA-N
Synonym 1-adamantanethiol, adamantanethiol, 1-adamantyl hydrosulfide, tricyclo 3.3.1.13,7 decane-1-thiol, tricyclo 3.3.1.1 3,7 decane-1-thiol, acmc-20aoyp, 1-admsh, pubchem21526, 1-adsh
PubChem CID 99730
IUPAC Name adamantane-1-thiol
SMILES C1C2CC3CC1CC(C2)(C3)S

Specifications

Melting Point 99°C to 106°C
Quantity 1 g
UN Number UN2811
Sensitivity Hygroscopic
Formula Weight 168.3
Percent Purity 98%
Chemical Name or Material 1-Mercaptoadamantane
Hazard Category H301-H319
Hazard Statement GHS H Statement
H301-H319
Toxic if swallowed.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P280i-P301+P310-P305+P351+P338-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.