Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methanesulfonyl-2-imidazolidinone 98.0+%, TCI America™
SDP

Catalog No. M255325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M255325G 25 g
M25535G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M255325G Supplier TCI America Supplier No. M255325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 41730-79-4
Molecular Formula C4H8N2O3S
Molecular Weight (g/mol) 164.179
MDL Number MFCD07787278
InChI Key WTCUHYRXLIHSLY-UHFFFAOYSA-N
PubChem CID 3016296
IUPAC Name 1-methylsulfonylimidazolidin-2-one
SMILES CS(=O)(=O)N1CCNC1=O

Specifications

Melting Point 193°C
Color White-Yellow
Quantity 25 g
Formula Weight 164.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Methanesulfonyl-2-imidazolidinone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.