Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methoxy-1-trimethylsilyloxypropene 95.0+%, TCI America™
SDP

Catalog No. M11991G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M11991G 1 g
M11995G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M11991G Supplier TCI America Supplier No. M11991G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 34880-70-1
Molecular Formula C7H16O2Si
Molecular Weight (g/mol) 160.288
MDL Number MFCD00671554
InChI Key RTHAKVSHSCPUGI-VOTSOKGWSA-N
Synonym [(1-Methoxy-1-propenyl)oxy]trimethylsilane
PubChem CID 10855775
IUPAC Name [(E)-1-methoxyprop-1-enoxy]-trimethylsilane
SMILES CC=C(OC)O[Si](C)(C)C

Specifications

Color Colorless
Boiling Point 52°C
Quantity 1 g
UN Number 1993
Formula Weight 160.29
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Methoxy-1-trimethylsilyloxypropene
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.