Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methoxy-2-butanol, Thermo Scientific Chemicals

Catalog No. AC466370250
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC466370250 25 mL
AC466370050 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC466370250 Supplier Thermo Scientific Chemicals Supplier No. 466370250
  • $123.00
    $76.65 / Each of 1

    Save $46.35

    38% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 53778-73-7 | C5H12O2 | 104.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53778-73-7
Molecular Formula C5H12O2
Molecular Weight (g/mol) 104.15
InChI Key CSZZMFWKAQEMPB-UHFFFAOYNA-N
IUPAC Name 1-methoxybutan-2-ol
SMILES CCC(O)COC

Specifications

Alpha Vector METHOXYBUTANOL 010
Color Colorless
Density 0.91
Boiling Point 135°C
Flash Point 39°C
Infrared Spectrum Conforms
Packaging glass bottle
Refractive Index 1.4090 to 1.4130 (20°C, 589 nm)
Quantity 25 mL
UN Number 1987
Formula Weight 104.15
Physical Form Liquid
Chemical Name or Material 1-Methoxy-2-butanol
Show More Show Less
Hazard Category GHS02: Flammable
ShelfLife 3 years
storageNote1 normal conditions

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.