Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methoxybicyclo[2.2.2]oct-5-ene-2-carbonitrile, Thermo Scientific™

Catalog No. AC202790250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC202790250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC202790250 Supplier Thermo Scientific Chemicals Supplier No. 202790250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38258-92-3
Molecular Formula C10H13NO
Molecular Weight (g/mol) 163.22
InChI Key XQFSLPUVWRXWSQ-UHFFFAOYSA-N
Synonym 1-methoxybicyclo 2.2.2 oct-5-ene-2-carbonitrile, 4-methoxybicyclo 2.2.2 oct-2-ene-5-carbonitrile, bicyclo 2.2.2 oct-5-ene-2-carbonitrile,1-methoxy
PubChem CID 2724147
IUPAC Name 4-methoxybicyclo[2.2.2]oct-2-ene-5-carbonitrile
SMILES COC12CCC(CC1C#N)C=C2

Specifications

Density 1.05
Quantity 25 g
Formula Weight 163.22
Chemical Name or Material 1-Methoxybicyclo[2.2.2]oct-5-ene-2-carbonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.