Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(Methoxymethyl)-1H-benzotriazole 98.0+%, TCI America™
SDP

Catalog No. M12761G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M12761G 1 g
M12765G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M12761G Supplier TCI America Supplier No. M12761G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 71878-80-3
Molecular Formula C8H9N3O
Molecular Weight (g/mol) 163.18
MDL Number MFCD00216630
InChI Key QEIXBXXKTUNWDK-UHFFFAOYSA-N
PubChem CID 542390
IUPAC Name 1-(methoxymethyl)-1H-1,2,3-benzotriazole
SMILES COCN1N=NC2=CC=CC=C12

Specifications

Melting Point 40°C
Color White-Yellow
Quantity 1 g
Formula Weight 163.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-(Methoxymethyl)-1H-benzotriazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.