Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl-1,4-cyclohexadiene 95.0+%, TCI America™
SDP

Catalog No. M048625ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M048625ML 25 mL
M04865ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M048625ML Supplier TCI America Supplier No. M048625ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4313-57-9
Molecular Formula C7H10
Molecular Weight (g/mol) 94.16
MDL Number MFCD00001538
InChI Key QDXQAOGNBCOEQX-UHFFFAOYSA-N
Synonym 1-methyl-1,4-cyclohexadiene, 2,5-dihydrotoluene, 1,4-cyclohexadiene, 1-methyl, 1, 1-methyl, acmc-1ast4, 1-methyl1,4-cyclohexadiene, 1-methylcyclohexane-1,4-diene, 1-methyl-1,4-cyclohexadiene, stabilized
PubChem CID 78006
IUPAC Name 1-methylcyclohexa-1,4-diene
SMILES CC1=CCC=CC1

Specifications

Color Yellow
Boiling Point 115°C
Quantity 25 mL
UN Number 3295
Formula Weight 94.16
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Methyl-1,4-cyclohexadiene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.