Learn More
Sigma Organic Chemistry 1-Methyl-1H-indazole-4- | 1G | 1053655-56-3 | MFCD10699167
Supplier: Sigma Organic Chemistry SYX001351G

1-Methyl-1H-indazole-4- , 1G
About this item:
CAS #: 1053655-56-3
MDL #: MFCD10699167
Chemical Formula: C9H8N2O
Molecular Weight: 160.17
UNSPSC Code: 12352200
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.