Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl-1H-indole-6-carbaldehyde, 97%, Thermo Scientific™

Catalog No. CC44204DA
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC44204DA 1 g Amber Glass Bottle
CC44204CB 250 mg Amber Glass Bottle
CC44204DE 5 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC44204DA Supplier Thermo Scientific Chemicals Supplier No. CC44204DA
Add to Cart

Chemical Identifiers

CAS 21005-45-8
Molecular Formula C10H9NO
Molecular Weight (g/mol) 159.19
MDL Number MFCD08690251
InChI Key QGAKKGIWURKKHV-UHFFFAOYSA-N
Synonym 1-methyl-1h-indole-6-carbaldehyde, 1h-indole-6-carboxaldehyde, 1-methyl, 1h-indole-6-carboxaldehyde,1-methyl, 6-formy-1-methylindole, 6-formyl-1-methylindole, 1-methyl-1h-indole-6-carboxaldehyde
PubChem CID 14835679
IUPAC Name 1-methylindole-6-carbaldehyde
SMILES CN1C=CC2=CC=C(C=O)C=C12

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 159.19
Percent Purity 97%
Chemical Name or Material 1-Methyl-1H-indole-6-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.