Learn More
Sigma Organic Chemistry 1-Methyl-1H-pyrazole-3 | 1G | MFCD22666523
Supplier: Sigma Organic Chemistry CBR017181G

1-Methyl-1H-pyrazole-3 , 1G
About this Item:
MDL #: MFCD22666523
Molecular Weight: 185.06
UNSPSC Code: 12352200
Chemical Formula: C4H10Cl2N4
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.