Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl-2-pyrrolidinone, ≥99.9% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019082 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9082 10 mL
11-101-9923 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 11-101-9082 Supplier MilliporeSigma Supelco Supplier No. 7876910MLF
Add to Cart
Add to Cart
This item is not returnable. View return policy

1-Methyl-2-pyrrolidinone is a polar solvent widely used in organic chemistry and polymer chemistry. On a large scale, it finds applications in the recovery and purification of acetylenes, olefins, and diolefins, gas purification, and aromatics extraction from feedstocks.

Featured Industries: Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Ignition Point 518°F
CAS 872-50-4
Vapor Pressure 0.29 mmHg (20°C)
Vapor Density 3.4 (vs. air)
Density 1.028 g/mL (at 25°C)
Flash Point 91°C
Physical Form Neat
Linear Formula C5H9 NO
MDL Number MFCD00003193
Melting Point -24°C
Boiling Point 202°C; 81°C to 82°C (10 mmHg)
Synonym 1-Methyl-2-pyrrolidone; N-Methyl-2-pyrrolidone; NMP
Type Analytical Standard
Quantity 10 mL
Formula Weight 99.13
Percent Purity ≥99.9% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.