Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl-2-pyrrolidinone, Biotech Grade, ≥99.7%, Solstice

Catalog No. 60046992 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2 L
2.5 L
4 x 2 L
Packaging:
Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-046-992 4 x 2 L Glass Bottle
60-046-990 2.5 L Glass Bottle
60-046-989 1 L Glass Bottle
60-046-991 2 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 60-046-992 Supplier Solstice Supplier No. 4944964X2L

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Biotech. grade,≥99.7%

  • Solstice developed the first high-purity solvents more than 50 years ago
  • As new applications and techniques emerge, Solstice continues to develop products to meet the most demanding requirements for laboratories and pharmaceutical production
  • Our packaging solutions, ranging from lab scale through production, provide our customers with added value, increased efficiency, and enhanced safety.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 872-50-4
Molecular Formula C5H9NO
Molecular Weight (g/mol) 99.133
MDL Number MFCD00003193
InChI Key SECXISVLQFMRJM-UHFFFAOYSA-N
Synonym 1-methyl-2-pyrrolidinone, n-methyl-2-pyrrolidone, n-methylpyrrolidone, 1-methyl-2-pyrrolidone, methylpyrrolidone, m-pyrol, n-methyl-2-pyrrolidinone, n-methylpyrrolidinone, 1-methylpyrrolidinone, 2-pyrrolidinone, 1-methyl
PubChem CID 13387
ChEBI CHEBI:7307
IUPAC Name 1-methylpyrrolidin-2-one
SMILES CN1CCCC1=O

Specifications

pH 8.5 to 10 (at 20°C)
Melting Point -24°C
Density 1.028g/mL at 25°C
Quantity 4 x 2 L
Explosion Limit 0.095
Flash Point 91°C (195.8°F)
Ignition Point 518°F
Odor Amine like
Packaging Glass Bottle
Physical Form Liquid
Vapor Pressure 0.99mmHg (40°C), 0.29mmHg (20°C)
Beilstein 106420
Boiling Point 204°C
Formula Weight 99.13g/mol
Percent Purity ≥99.7%
Chemical Name or Material 1-Methyl-2-pyrrolidinone
Show More Show Less
Hazard Statement P201-P280-P305 + P351 + P338-P308 + P313
RTECSNumber 212-828-1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.