Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl-3-n-octylimidazolium Chloride 98.0+%, TCI America™
SDP

Catalog No. M206225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M206225G 25 g
M20625G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M206225G Supplier TCI America Supplier No. M206225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 64697-40-1
Molecular Formula C12H23ClN2
Molecular Weight (g/mol) 230.78
MDL Number MFCD03095432
InChI Key OXFBEEDAZHXDHB-UHFFFAOYSA-M
Synonym 1-methyl-3-n-octylimidazolium chloride, 1-methyl-3-octylimidazolium chloride, 1-octyl-3-methylimidazolium chloride, 3-methyl-1-octylimidazolium chloride, omimcl, acmc-20aj1j, dsstox_cid_27849, dsstox_rid_82602, dsstox_gsid_47873, ksc491c7j
PubChem CID 2734223
ChEBI CHEBI:61342
IUPAC Name 3-methyl-1-octyl-1H-imidazol-3-ium chloride
SMILES [Cl-].CCCCCCCCN1C=C[N+](C)=C1

Specifications

Melting Point 12°C
Quantity 25 g
Formula Weight 230.78
Percent Purity ≥98.0% (HPLC,T)
Physical Form Liquid
Chemical Name or Material 1-Methyl-3-n-octylimidazolium Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.