Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl-3-n-octylimidazolium Hexafluorophosphate 98.0+%, TCI America™
SDP

Catalog No. M206325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M206325G 25 g
M20635G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M206325G Supplier TCI America Supplier No. M206325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 304680-36-2
Molecular Formula C12H23F6N2P
Molecular Weight (g/mol) 340.29
MDL Number MFCD03427617
InChI Key GRCIJNHHTXBJAK-UHFFFAOYSA-N
Synonym 1-methyl-3-n-octylimidazolium hexafluorophosphate, 3-methyl-1-octylimidazolium hexafluorophosphate, 1-methyl-3-octylimidazolium hexafluorophosphate, 1-octyl-3-methylimidazolium hexafluorophosphate, 1-methyl-3-n-octylimidazoliumhexafluorophosphate, acmc-20aj2z, dsstox_cid_27928, dsstox_rid_82681, dsstox_gsid_47952, ksc222c5l
PubChem CID 2734243
IUPAC Name 3-methyl-1-octyl-1H-imidazol-3-ium; hexafluoro-λ⁵-phosphanuide
SMILES F[P-](F)(F)(F)(F)F.CCCCCCCCN1C=C[N+](C)=C1

Specifications

Color Yellow
Quantity 25 g
Formula Weight 340.29
Percent Purity ≥98.0% (HPLC,N)
Physical Form Liquid
Chemical Name or Material 1-Methyl-3-n-octylimidazolium Hexafluorophosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.