Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl-3-pentylimidazolium Bromide 98.0+%, TCI America™
SDP

Catalog No. M30375G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M30375G 5 g
M303725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M30375G Supplier TCI America Supplier No. M30375G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 343851-31-0
Molecular Formula C9H17BrN2
Molecular Weight (g/mol) 233.153
MDL Number MFCD19382537
InChI Key PXFKRXXEFJDOMO-UHFFFAOYSA-M
Synonym 1-Methyl-3-pentyl-1H-imidazol-3-ium Bromide
PubChem CID 10537569
IUPAC Name 1-methyl-3-pentylimidazol-1-ium;bromide
SMILES CCCCCN1C=C[N+](=C1)C.[Br-]

Specifications

Quantity 5 g
Formula Weight 233.15
Percent Purity ≥98.0% (HPLC)
Physical Form Liquid
Chemical Name or Material 1-Methyl-3-pentylimidazolium Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.