Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl-3-pentylimidazolium Tetrafluoroborate, TCI America™
SDP

Catalog No. M303825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M303825G 25 g
M30385G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M303825G Supplier TCI America Supplier No. M303825G

Chemical Identifiers

CAS 244193-49-5
Molecular Formula C9H17BF4N2
MDL Number MFCD19382533

Specifications

Color Yellow
Quantity 25 g
UN Number 1760
Formula Weight 240.05
Percent Purity ≥98.0% (N)
Physical Form Liquid
Chemical Name or Material 1-Methyl-3-pentylimidazolium Tetrafluoroborate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.