Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl-3-propylimidazolium Iodide 97.0+%, TCI America™
SDP

Catalog No. M144025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M144025G 25 g
M14405G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M144025G Supplier TCI America Supplier No. M144025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 119171-18-5
Molecular Formula C7H13IN2
Molecular Weight (g/mol) 252.10
MDL Number MFCD08276385
InChI Key IVCMUVGRRDWTDK-UHFFFAOYSA-M
Synonym 1-methyl-3-propylimidazolium iodide, 1-propyl-3-methylimidazolium iodide, 1-methyl-3-n-propylimidazolium iodide, 1-methyl-3-propylimidazol-1-ium iodide, acmc-1c1fw, dsstox_cid_29063, dsstox_rid_83282, dsstox_gsid_49207, ksc491c6t, 1-methyl-3-propylimidazoliumiodide
PubChem CID 12971008
IUPAC Name 3-methyl-1-propyl-1H-imidazol-3-ium iodide
SMILES [I-].CCCN1C=C[N+](C)=C1

Specifications

Quantity 25 g
Formula Weight 252.10
Percent Purity ≥97.0% (HPLC,T)
Physical Form Liquid
Chemical Name or Material 1-Methyl-3-propylimidazolium Iodide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.