Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl-3-pyrrolidinol, 97%

Catalog No. AC127620010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC127620010 1 g Glass bottle
AC127620050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC127620010 Supplier Thermo Scientific Chemicals Supplier No. 127620010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 13220-33-2 | C5H11NO | 101.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13220-33-2
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.15
MDL Number MFCD00003176
InChI Key FLVFPAIGVBQGET-UHFFFAOYSA-N
Synonym 1-methyl-3-pyrrolidinol, 3-hydroxy-1-methylpyrrolidine, n-methyl-3-pyrrolidinol, 3-pyrrolidinol, 1-methyl, 3-hydroxy-n-methylpyrrolidine, n-methyl-3-hydroxypyrrolidine, 1-methyl-3-hydroxypyrrolidine, pubchem14117, l-methyl-3-pyrrolidinol, 1-methyl-pyrrolidin-3ol
PubChem CID 93074
IUPAC Name 1-methylpyrrolidin-3-ol
SMILES CN1CCC(C1)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Density 0.9210g/mL
Boiling Point 50.0°C to 52.0°C (1.0 mmHg)
Flash Point 71°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Beilstein 21, II, 3
Specific Gravity 0.921
Formula Weight 101.15
Percent Purity 97%
Chemical Name or Material 1-Methyl-3-pyrrolidinol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
EINECSNumber 236-189-3

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.