Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl-4-[1-(4-piperidyl)-4-piperidyl]piperazine 98.0+%, TCI America™
SDP

Catalog No. M219725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M219725G 25 g
M21975G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M219725G Supplier TCI America Supplier No. M219725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1629218-99-0
Molecular Formula C15H30N4
Molecular Weight (g/mol) 266.43
MDL Number MFCD20257913
InChI Key VVNRVKMOTTTYKS-UHFFFAOYSA-N
PubChem CID 53384439
IUPAC Name 4-(4-methylpiperazin-1-yl)-1,4'-bipiperidine
SMILES CN1CCN(CC1)C1CCN(CC1)C1CCNCC1

Specifications

Melting Point 89°C
Color White-Yellow
Quantity 25 g
Synonym 1-Methyl-4-(1,4′-bipiperidin-4-yl)piperazine
Formula Weight 266.43
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Methyl-4-[1-(4-piperidyl)-4-piperidyl]piperazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.