Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl-4-(4-nitrophenyl)piperazine, 97%, Thermo Scientific™

Catalog No. AAH6366503
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6366503 1 g
AAH63665MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6366503 Supplier Thermo Scientific Chemicals Supplier No. H6366503

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16155-03-6
Molecular Formula C11H15N3O2
Molecular Weight (g/mol) 221.26
MDL Number MFCD00101003
InChI Key GZNDUKANJZIZOT-UHFFFAOYSA-N
Synonym 1-methyl-4-4-nitrophenyl piperazine, 1-methyl-4-4-nitro-phenyl-piperazine, 4-methyl-1-4-nitrophenyl piperazine, acmc-1bvu1, cambridge id 5139345, 1-methyl-4-p-nitrophenyl piperazine, 1-4-nitrophenyl-4-methylpiperazine, 1-methyl-4-4-nitrophenyl piperizine, n-4-nitrophenyl-n'-methylpiperazine, 4-methyl-1-4-nitro-phenyl-piperazine
PubChem CID 2825198
IUPAC Name 1-methyl-4-(4-nitrophenyl)piperazine
SMILES CN1CCN(CC1)C2=CC=C(C=C2)[N+](=O)[O-]

Specifications

Quantity 1 g
Formula Weight 221.26
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-Methyl-4-(4-nitrophenyl)piperazine
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.