Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl-4-(methylamino)piperidine 97.0+%, TCI America™
SDP

Catalog No. M25661G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M25661G 1 g
M2566200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M25661G Supplier TCI America Supplier No. M25661G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 73579-08-5
Molecular Formula C7H16N2
Molecular Weight (g/mol) 128.219
MDL Number MFCD00006498
InChI Key XRYGCVVVDCEPRL-UHFFFAOYSA-N
PubChem CID 566323
IUPAC Name N,1-dimethylpiperidin-4-amine
SMILES CNC1CCN(CC1)C

Specifications

Boiling Point 53°C
Quantity 1 g
UN Number 2920
Formula Weight 128.22
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-Methyl-4-(methylamino)piperidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.