Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl-4-phenyl-1,2,3,6-tetrahydropyridine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. M2690100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M2690100MG 100 mg
M269025MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M2690100MG Supplier TCI America Supplier No. M2690100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 23007-85-4
Molecular Formula C12H16ClN
Molecular Weight (g/mol) 209.72
MDL Number MFCD00012790
InChI Key KOWJANGMTAZWDT-UHFFFAOYSA-N
Synonym MPTP Hydrochloride
PubChem CID 161406
IUPAC Name hydrogen 1-methyl-4-phenyl-1,2,3,6-tetrahydropyridine chloride
SMILES [H+].[Cl-].CN1CCC(=CC1)C1=CC=CC=C1

Specifications

Melting Point 250°C
Color White-Yellow
Quantity 100 mg
UN Number 2811
Formula Weight 209.72
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Methyl-4-phenyl-1,2,3,6-tetrahydropyridine Hydrochloride
RTECSNumber UT8358950
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.