Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl-4-piperidone, 98%

Catalog No. AAA1335218
Change view
Click to view available options
Quantity:
50 g
100 g
250 g
1000 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1335218 50 g
AAA1335222 100 g
AAA1335230 250 g
AAA133520B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1335218 Supplier Thermo Scientific Chemicals Supplier No. A1335218
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 1445-73-4 | C6H11NO | 113.16 g/mol

1-Methyl-4-piperidone, are widely used in artificial fibre industry. They are polymerizable and used as nylon precursors. They have excellent applications as solvents and as intermediates for organic synthesis. They are used in the aqueous carrier medium and amide penetrants in inks and water soluble paints.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Methyl-4-piperidone, are widely used in artificial fibre industry. They are polymerizable and used as nylon precursors. They have excellent applications as solvents and as intermediates for organic synthesis. They are used in the aqueous carrier medium and amide penetrants in inks and water soluble paints.

Solubility
It is miscible with water

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from strong oxidizing agents, strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1445-73-4
Molecular Formula C6H11NO
Molecular Weight (g/mol) 113.16
MDL Number MFCD00006191
InChI Key HUUPVABNAQUEJW-UHFFFAOYSA-N
Synonym 1-methyl-4-piperidone, n-methyl-4-piperidone, 1-methyl-4-piperidinone, 4-piperidinone, 1-methyl, 4-piperidone, 1-methyl, n-methyl-4-piperidinone, 1-methyl-4-piperidione, n-methylpiperidine-4-one, 1-methyl-4-oxopiperidine, 1-methylpiperidine-4-one
PubChem CID 74049
IUPAC Name 1-methylpiperidin-4-one
SMILES CN1CCC(=O)CC1

Specifications

Density 0.973
pH 12
Boiling Point 55°C to 58°C (10 mmHg)
Flash Point 60°C (140°F)
Odor Amine-like
Refractive Index 1.461
Quantity 50 g
UN Number UN2733
Beilstein 106924
Sensitivity Air sensitive
Solubility Information It is miscible with water
Formula Weight 113.16
Percent Purity 98%
Chemical Name or Material 1-Methyl-4-piperidone
Show More Show Less
Hazard Category H226-H314-H335
Hazard Statement GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
EINECSNumber 215-895-5
TSCA No
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.