Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl-4-piperidone 98.0+%, TCI America™
SDP

Catalog No. M053025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M053025G 25 g
M0530500G 500 g
M0530100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M053025G Supplier TCI America Supplier No. M053025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1445-73-4
Molecular Formula C6H11NO
Molecular Weight (g/mol) 113.16
MDL Number MFCD00006191
InChI Key HUUPVABNAQUEJW-UHFFFAOYSA-N
Synonym 1-methyl-4-piperidone, n-methyl-4-piperidone, 1-methyl-4-piperidinone, 4-piperidinone, 1-methyl, 4-piperidone, 1-methyl, n-methyl-4-piperidinone, 1-methyl-4-piperidione, n-methylpiperidine-4-one, 1-methyl-4-oxopiperidine, 1-methylpiperidine-4-one
PubChem CID 74049
IUPAC Name 1-methylpiperidin-4-one
SMILES CN1CCC(=O)CC1

Specifications

Color Yellow
Boiling Point 72°C
Quantity 25 g
UN Number 1224
Formula Weight 113.16
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-Methyl-4-piperidone
EINECSNumber (9)-2198
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.