Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methyl-5-mercaptotetrazole, Sodium Salt,dihydrate 98%, Thermo Scientific™

Catalog No. AC134691000
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC134691000 100 g
AC134690250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC134691000 Supplier Thermo Scientific Chemicals Supplier No. 134691000

Item Discontinued This product is either new or inactive, and cannot be ordered online.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51138-06-8
Molecular Formula C2H4N4NaS
Molecular Weight (g/mol) 139.132
MDL Number MFCD00149121
InChI Key KPTNUCBVMWICQI-UHFFFAOYSA-N
Synonym sodium 1-methyl-1h-tetrazole-5-thiolate, potassium 1-methyl-1,2,3,4-tetrazol-5-yl sulfanide, cefminox sodium impurity 2, sodium 1-methyltetrazole-5-thiolate, 1-methyl-5-mercaptotetrazole,sodiumsalt, 1-methyl-5-mercaptotetrazole, sodium, 1-methyl-5-sodiosulfanyl-1,2,3,4-tetrazole, sodium 1-methyl-1,2,3,4-tetrazol-5-yl sulfanide
PubChem CID 67016449
IUPAC Name 1-methyl-2H-tetrazole-5-thione;sodium
SMILES CN1C(=S)N=NN1.[Na]

Specifications

CAS Min % 97.5
CAS Max % 100.0
Quantity 100 g
Formula Weight 174.15
Chemical Name or Material 1-Methyl-5-mercaptotetrazole, sodium salt, dihydrate, 98%
Hazard Category GHS Signal Word: Warning
EINECSNumber 257-006-3
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.