Learn More
Bio-techne Sales Corp 1% Methyl Cellulose Solution, 100 mL
SDP
Catalog No. NC9874022 Shop All Novus Biologicals Products
Encompass
This solution is used to prepare samples for the cIEF applications. The expiration date for this kit is 12 months from date of manufacture. For use with Maurice, Maurice C., iCE3, and iCE280 systems only.
Catalog No. NC9874022 Supplier Bio-techne Sales Corp Supplier No. 101876
Add to Cart
Add to Cart
This solution is used to prepare samples for the cIEF applications. The expiration date for this kit is 12 months from date of manufacture. For use with Maurice, Maurice C., iCE3, and iCE280 systems only.