Learn More
TARGETMOL CHEMICALS INC 1-METHYL-L-HISTIDINE 50MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T493450MG
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg and bulk. Please contact Fisher for quotes. 1-Methyl-L-histidine (3-(1-Methylimidazol-4-yl)-L-alanine) is a natural but non-proteinogenic amino acid; employed as index of muscle protein breakdown. purity: 99%
Catalog No. 50-257-6730
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.