Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methylbenzimidazole 98.0+%, TCI America™
SDP

Catalog No. M217325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M217325G 25 g
M21735G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M217325G Supplier TCI America Supplier No. M217325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1632-83-3
Molecular Formula C8H8N2
Molecular Weight (g/mol) 132.17
MDL Number MFCD00192275
InChI Key FGYADSCZTQOAFK-UHFFFAOYSA-N
Synonym 1-methyl-1h-benzimidazole, 1h-benzimidazole, 1-methyl, methylbenzimidazole, unii-12lw89n19y, 1-methyl-1h-benzo d imidazole, 1h-benzimidazole, methyl, 1-methyl-1,3-benzodiazole, 1-methyl-1h-1,3-benzodiazole, pubchem7613, 1-methylbenzimidazole
PubChem CID 95890
IUPAC Name 1-methyl-1H-1,3-benzodiazole
SMILES CN1C=NC2=CC=CC=C12

Specifications

Melting Point 59°C
Boiling Point 154°C
Quantity 25 g
Formula Weight 132.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Methylbenzimidazole
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.