Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methylindole-2-carboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. M15191G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M15191G 1 g
M15195G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M15191G Supplier TCI America Supplier No. M15191G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 16136-58-6
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.187
MDL Number MFCD00005801
InChI Key MAHAMBLNIDMREX-UHFFFAOYSA-N
Synonym 1-methyl-1h-indole-2-carboxylic acid, 1h-indole-2-carboxylic acid, 1-methyl, 1-methyl-2-indolecarboxylic acid, indole-2-carboxylic acid, 1-methyl, chembl36834, n-methylindolcarbonsaure, pubchem20609, acmc-1bpru, 5-22-03-00015 beilstein handbook reference, mahamblnidmrex-uhfffaoysa
PubChem CID 27695
IUPAC Name 1-methylindole-2-carboxylic acid
SMILES CN1C2=CC=CC=C2C=C1C(=O)O

Specifications

Melting Point 212°C
Quantity 1 g
Formula Weight 175.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Methylindole-2-carboxylic Acid
RTECSNumber NL6012400
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.