Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methylindole, 98+%

Catalog No. p-3779124
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC186300050 5 g Glass bottle
AC186300250 25 g Glass bottle
AC186301000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC186300050 Supplier Thermo Scientific Chemicals Supplier No. 186300050
Only null left
Add to Cart

CAS: 603-76-9 | C9H9N | 131.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 603-76-9
Molecular Formula C9H9N
Molecular Weight (g/mol) 131.18
MDL Number MFCD00005800
InChI Key BLRHMMGNCXNXJL-UHFFFAOYSA-N
Synonym 1-methyl-1h-indole, n-methylindole, 1h-indole, 1-methyl, indole, 1-methyl, 1-methylindol, methylindole, unii-8h698roj5f, n-, chembl19912, n-methylindol
PubChem CID 11781
IUPAC Name 1-methylindole
SMILES CN1C=CC2=CC=CC=C21

Specifications

CAS Min % 98.0
CAS Max % 100.0
Color Colorless to White
Density 1.0200g/mL
Boiling Point 236°C to 239°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.6060 to 1.608
Quantity 5 g
Specific Gravity 1.02
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol,ethyl ether,benzene
Formula Weight 131.18
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 1-Methylindole
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 210-057-5
Recommended Storage This product darkens on exposure to light

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.