Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methylnaphthalene, EPA Individual Analyte, 1000 μg/mL in Methanol, 1 mL

Catalog No. USEPA1225
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-EPA-1225 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-EPA-1225 Supplier Agilent Technologies Supplier No. EPA12251
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 90-12-0 , 67-56-1
Molecular Formula C11H10
Molecular Weight (g/mol) 142.201
InChI Key QPUYECUOLPXSFR-UHFFFAOYSA-N
PubChem CID 7002
ChEBI CHEBI:50717
IUPAC Name 1-methylnaphthalene
SMILES CC1=CC=CC2=CC=CC=C12

Specifications

CAS Max % 99.87, 0.12
Analyte or Component Names methanol,1,2:3
Element methanol,1,2:3
Solvent or Matrix Methanol
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) 1000μg/mL
Chemical Name or Material 1-Methylnaphthalene Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.