Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ 1-Methylpsilocin

Catalog No. 301750
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-175-0 50 mg
30-171-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 30-175-0 Supplier Tocris Bioscience™ Supplier No. 3017/50
This item is not returnable. View return policy

Potent and selective 5-HT2C agonist

Potent and selective 5-HT2C agonist (IC50 values are 12 and 633 nM for 5-HT2C and 5-HT2A receptors respectively). Displays high affinity for the 5-HT2B receptor (Ki = 38 nM) but acts as an inverse agonist. Active in vivo; inhibits scratching in a mouse model of OCD.

Chemical Identifiers

CAS 1465-16-3
Molecular Formula C13H18N2O

Specifications

Quantity 50 mg
Formula Weight 218.29
Purity >99%
Molecular Weight (g/mol) 218.3
Chemical Name or Material 1-Methylpsilocin
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.