Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methylpyrazole-5-carboxylic Acid 97.0+%, TCI America™
SDP

Catalog No. M24945G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M24945G 5 g
M24941G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M24945G Supplier TCI America Supplier No. M24945G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 16034-46-1
Molecular Formula C5H6N2O2
Molecular Weight (g/mol) 126.12
MDL Number MFCD00464253
InChI Key JREJQAWGQCMSIY-UHFFFAOYSA-N
Synonym 1-methyl-1h-pyrazole-5-carboxylic acid, 1-methylpyrazole-5-carboxylic acid, 2-methyl-2h-pyrazole-3-carboxylic acid, 1h-pyrazole-5-carboxylic acid, 1-methyl, 1-methyl-1h-pyrazole-5-carboxylicacid, 5-carboxy-1-methyl-1h-pyrazole, 1-methyl-5-pyrazolecarboxylic acid, 2h-pyrazole-3-carboxylic acid, 2-methyl, acmc-209dkx, ksc173g6t
PubChem CID 643158
ChEBI CHEBI:74737
IUPAC Name 1-methyl-1H-pyrazole-5-carboxylic acid
SMILES CN1N=CC=C1C(O)=O

Specifications

Melting Point 225°C
Color White
Quantity 5 g
Formula Weight 126.12
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Methylpyrazole-5-carboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.