Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methylpyrazole-5-methanol 98.0+%, TCI America™
SDP

Catalog No. M28311G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M28311G 1 g
M2831200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M28311G Supplier TCI America Supplier No. M28311G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 84547-61-5
Molecular Formula C5H8N2O
Molecular Weight (g/mol) 112.132
MDL Number MFCD08556301
InChI Key WQFOGLYQVFBDEY-UHFFFAOYSA-N
Synonym 1-methyl-1h-pyrazol-5-yl methanol, 5-hydroxymethyl-1-methyl-1h-pyrazole, 1h-pyrazole-5-methanol, 1-methyl, 2-methylpyrazol-3-yl methanol, 5-hydroxymethyl-1-methylpyrazole, 1h-pyrazole-5-methanol,1-methyl, 1-methylpyrazol-5-yl methan-1-ol, 1-methylpyrazole-5-methanol, 1-methyl-1h-pyrazol-5-yl ;methanol
PubChem CID 11815316
IUPAC Name (2-methylpyrazol-3-yl)methanol
SMILES CN1C(=CC=N1)CO

Specifications

Melting Point 58°C
Color White
Quantity 1 g
Formula Weight 112.13
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Methylpyrazole-5-methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.