Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methylpyridinium Chloride 98.0+%, TCI America™
SDP

Catalog No. M139725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M139725G 25 g
M13975G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M139725G Supplier TCI America Supplier No. M139725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7680-73-1
Molecular Formula C6H8ClN
Molecular Weight (g/mol) 129.59
MDL Number MFCD00137532
InChI Key QAIGYXWRIHZZAA-UHFFFAOYSA-M
PubChem CID 82116
IUPAC Name 1-methylpyridin-1-ium chloride
SMILES [Cl-].C[N+]1=CC=CC=C1

Specifications

Melting Point 144°C
Color White-Yellow
Quantity 25 g
UN Number 2811
Formula Weight 129.59
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Methylpyridinium Chloride
RTECSNumber UU5950000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.