Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-n-Butyl-3-methylimidazolium nitrate, 95%, Thermo Scientific™

Catalog No. AAH5980306
Change view
Click to view available options
Quantity:
5 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5980306 5 g
AAH5980318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5980306 Supplier Thermo Scientific Chemicals Supplier No. H5980306

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 179075-88-8
Molecular Formula C8H15N3O3
Molecular Weight (g/mol) 201.226
MDL Number MFCD08457696
InChI Key WPHIMOZSRUCGGU-UHFFFAOYSA-N
Synonym 1-butyl-3-methylimidazolium nitrate, 1-butyl-3-methyl-1h-imidazol-3-ium nitrate, bmim no3, c4mim nitrate, ksc171c6p, 1-butyl-3-methylimidazoliumnitrate, 1-butyl-3-methyl-imidazol-3-ium nitrate, 1-butyl-3-methylimidazolium nitrate hplc
PubChem CID 16211113
IUPAC Name 1-butyl-3-methylimidazol-3-ium;nitrate
SMILES CCCCN1C=C[N+](=C1)C.[N+](=O)([O-])[O-]

Specifications

Quantity 5 g
UN Number UN1479
Sensitivity Hygroscopic; May form a precipitate
Formula Weight 201.22
Percent Purity 95%
Chemical Name or Material 1-n-Butyl-3-methylimidazolium nitrate
Hazard Category H272-H315-H319-H335
Hazard Statement GHS H Statement
H272-H315-H319-H335
May intensify fire; oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.