Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Naphthol-3,6-disulfonic Acid, Disodium Salt (Tech.), Thermo Scientific™

Catalog No. AC199532500
Click to view available options
Quantity:
50 g
250 g

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20349-39-7
Molecular Formula C10H6Na2O7S2
Molecular Weight (g/mol) 348.251
InChI Key RJMDNLVCFLKDBY-UHFFFAOYSA-L
Synonym 1-naphthol-3,6-disulfonic acid disodium salt, 4-hydroxy-2,7-naphthalenedisulfonic acid disodium salt, 2,7-naphthalenedisulfonic acid, 4-hydroxy-, disodium salt, sodium 4-hydroxynaphthalene-2,7-disulfonate, 1-naphthol-3,6-disulfonic acid, sodium salt, disodium 4-hydroxynaphthalene-2,7-disulfonate, disodium 4-hydroxynaphthalene-2,7-disulphonate, 2,7-naphthalenedisulfonic acid, 4-hydroxy-, sodium salt 1:2, violet acid disodium salt, naphthol disulfonic acid sodium
PubChem CID 88502
IUPAC Name disodium;4-hydroxynaphthalene-2,7-disulfonate
SMILES C1=CC2=C(C=C(C=C2C=C1S(=O)(=O)[O-])S(=O)(=O)[O-])O.[Na+].[Na+]

Specifications

Color Gray-Pink
Infrared Spectrum Authentic
Linear Formula HOC10H5(SO3Na)2
Quantity 250 g
Beilstein 11, 227
Formula Weight 348.25
Percent Purity ≥40% (calculated with reference to the free acid (MW: 304))
Grade Technical
Physical Form Press Cake
Chemical Name or Material 1-Naphthol-3, 6-disulfonic acid, disodium salt, tech.
EINECSNumber 243-754-8
TSCA TSCA
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.