Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Naphthol-3,6-disulfonic Acid, Disodium Salt (Tech.), Thermo Scientific™

Catalog No. AC199530500
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC199530500 50 g
AC199532500 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC199530500 Supplier Thermo Scientific Chemicals Supplier No. 199530500

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20349-39-7
Molecular Formula C10H6Na2O7S2
Molecular Weight (g/mol) 348.251
InChI Key RJMDNLVCFLKDBY-UHFFFAOYSA-L
Synonym 1-naphthol-3,6-disulfonic acid disodium salt, 4-hydroxy-2,7-naphthalenedisulfonic acid disodium salt, 2,7-naphthalenedisulfonic acid, 4-hydroxy-, disodium salt, sodium 4-hydroxynaphthalene-2,7-disulfonate, 1-naphthol-3,6-disulfonic acid, sodium salt, disodium 4-hydroxynaphthalene-2,7-disulfonate, disodium 4-hydroxynaphthalene-2,7-disulphonate, 2,7-naphthalenedisulfonic acid, 4-hydroxy-, sodium salt 1:2, violet acid disodium salt, naphthol disulfonic acid sodium
PubChem CID 88502
IUPAC Name disodium;4-hydroxynaphthalene-2,7-disulfonate
SMILES C1=CC2=C(C=C(C=C2C=C1S(=O)(=O)[O-])S(=O)(=O)[O-])O.[Na+].[Na+]

Specifications

Color Gray-Pink
Infrared Spectrum Authentic
Linear Formula HOC10H5(SO3Na)2
Quantity 50 g
Beilstein 11, 227
Formula Weight 348.25
Percent Purity ≥40% (calculated with reference to the free acid (MW: 304))
Grade Technical
Physical Form Press Cake
Chemical Name or Material 1-Naphthol-3, 6-disulfonic acid, disodium salt, tech.
EINECSNumber 243-754-8
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.