Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Naphthol (alpha-Naphthol; 1-Hydroxynaphthalene), Reagents

Catalog No. 01337162
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-162 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-337-162 Supplier Reagents Holdings Llc Supplier No. C2252500100C2
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Naphthol (alpha-Naphthol; 1-Hydroxynaphthalene), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 90-15-3
Molecular Formula C10H8O
Molecular Weight (g/mol) 144.17
InChI Key KJCVRFUGPWSIIH-UHFFFAOYSA-N
Synonym 1-Naphthalenol
IUPAC Name naphthalen-1-ol
SMILES OC1=C2C=CC=CC2=CC=C1

Specifications

Boiling Range 288°C
Melting Point 96°C
Color White or off-White
Boiling Point 288°C
Melting Range 96°C
Packaging Glass Bottle
Quantity 100 g
Specific Gravity 1.0989
Grade Reagent
Physical Form Solid
Chemical Name or Material 1-Naphthol (alpha-Naphthol, 1-Hydroxynaphthalene)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.