CAS RN 90-15-3
1-Naphthol 99.0+%, TCI America™
CAS: 90-15-3 Molecular Formula: C10H8O Molecular Weight (g/mol): 144.173 MDL Number: MFCD00003930 InChI Key: KJCVRFUGPWSIIH-UHFFFAOYSA-N Synonym: 1-naphthol,1-naphthalenol,alpha-naphthol,1-hydroxynaphthalene,naphthol,fouramine ern,fourrine ern,tertral ern,furro er,basf ursol ern PubChem CID: 7005 ChEBI: CHEBI:10319 IUPAC Name: naphthalen-1-ol SMILES: C1=CC=C2C(=C1)C=CC=C2O
1-Naphthol, 99%
CAS: 90-15-3 Molecular Formula: C10H8O Molecular Weight (g/mol): 144.173 MDL Number: MFCD00003930 InChI Key: KJCVRFUGPWSIIH-UHFFFAOYSA-N Synonym: 1-naphthol,1-naphthalenol,alpha-naphthol,1-hydroxynaphthalene,naphthol,fouramine ern,fourrine ern,tertral ern,furro er,basf ursol ern PubChem CID: 7005 ChEBI: CHEBI:10319 IUPAC Name: naphthalen-1-ol SMILES: C1=CC=C2C(=C1)C=CC=C2O
1-Naphthol, 99+%
CAS: 90-15-3 Molecular Formula: C10H8O Molecular Weight (g/mol): 144.17 MDL Number: MFCD00003930 InChI Key: KJCVRFUGPWSIIH-UHFFFAOYSA-N Synonym: 1-naphthol,1-naphthalenol,alpha-naphthol,1-hydroxynaphthalene,naphthol,fouramine ern,fourrine ern,tertral ern,furro er,basf ursol ern PubChem CID: 7005 ChEBI: CHEBI:10319 IUPAC Name: naphthalen-1-ol SMILES: C1=CC=C2C(=C1)C=CC=C2O
1-Naphthol, Sigma-Aldrich
CAS: 90-15-3 Molecular Weight (g/mol): 144.17 g/mol
1-Naphthol 98.0+%, TCI America™
CAS: 90-15-3 Molecular Formula: C10H8O Molecular Weight (g/mol): 144.173 MDL Number: MFCD00003930 InChI Key: KJCVRFUGPWSIIH-UHFFFAOYSA-N Synonym: 1-naphthol,1-naphthalenol,alpha-naphthol,1-hydroxynaphthalene,naphthol,fouramine ern,fourrine ern,tertral ern,furro er,basf ursol ern PubChem CID: 7005 ChEBI: CHEBI:10319 IUPAC Name: naphthalen-1-ol SMILES: C1=CC=C2C(=C1)C=CC=C2O
1-Naphthol (alpha-Naphthol; 1-Hydroxynaphthalene), Reagents
CAS: 90-15-3 Molecular Formula: C10H8O Molecular Weight (g/mol): 144.17 InChI Key: KJCVRFUGPWSIIH-UHFFFAOYSA-N Synonym: 1-Naphthalenol IUPAC Name: naphthalen-1-ol SMILES: OC1=C2C=CC=CC2=CC=C1
Sigma Aldrich L-(-)-Malic Acid Disodium Salt
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich 1-Naphthol
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich 1-Naphthol
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Fisher Science Education™ Molisch Reagent
CAS: 90-15-3 InChI Key: KJCVRFUGPWSIIH-UHFFFAOYSA-N Synonym: dihydrogen oxide,dihydrogen monoxide PubChem CID: 7005 ChEBI: CHEBI:10319
1-Naphthol, SPEX CertiPrep™
CAS: 90-15-3 Molecular Formula: C10H8O Molecular Weight (g/mol): 144.173 InChI Key: KJCVRFUGPWSIIH-UHFFFAOYSA-N PubChem CID: 7005 ChEBI: CHEBI:10319 IUPAC Name: naphthalen-1-ol SMILES: C1=CC=C2C(=C1)C=CC=C2O